Search
Search
| Gene Symbol: Cfap99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770377.1 | XP_008768599.1 | 16 | 2684 | 92 | |
| XM_008770378.1 | XP_008768600.1 | 17 | 2795 | 92 | ||
| XM_008770379.1 | XP_008768601.1 | 16 | 2707 | 92 | ||
| XM_008770380.1 | XP_008768602.1 | 14 | 6364 | 92 | ||
| XM_008770381.1 | XP_008768603.1 | 15 | 3372 | 92 | ||
Set Membership: |
|