Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn07311037_m1
          See other TNFRSF11B GE Assays ›
          Gene Symbol:
          Tnfrsf11b
          Gene Name:
          tumor necrosis factor receptor superfamily, member 11b
          Gene Aliases:
          Opg
          Chromosome Location:
          Chr.7: 93798580 - 93826586 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn07311037_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf11b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012870.2 NP_037002.1 4 - 5 917 73
          GenBank mRNA BC081830.1 - 4 - 5 917 7096099 73
          U94330.1 - 4 - 5 936 73

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf11b

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          signal transduction
          response to nutrient
          response to inorganic substance
          extracellular matrix organization
          response to magnesium ion
          negative regulation of odontogenesis of dentin-containing tooth
          response to drug
          response to estrogen
          negative regulation of bone resorption
          response to arsenic-containing substance
          molecular_function
          cytokine activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P458021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P458021 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P458021 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P458021 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P458021 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73131 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7445 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903730 - -
          Affymetrix Rat Gene 1.0 ST Array 10903728 - -
          Affymetrix Rat Gene 1.0 ST Array 10903729 - -
          Affymetrix Rat Gene 1.0 ST Array 10903726 - -
          Affymetrix Rat Gene 1.0 ST Array 10903727 - -
          Affymetrix Rat Gene 1.1 ST Array 10903725 - -
          Rat Genome 230 2.0 Array 1369407_PM_at - -
          Rat Genome 230 2.0 Array 1369407_at - -
          Affymetrix HT Rat Focus Array 1369407_PM_at - -
          Affymetrix HT Rat Focus Array 1369407_at - -
          Affymetrix Rat Clariom S Assay PSR0700024004.rn.1 TC0700003139.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024000.rn.1 TC0700003139.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024002.rn.1 TC0700003139.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024001.rn.1 TC0700003139.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024003.rn.1 TC0700003139.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024005.rn.1 TC0700003139.rn.1 RTA TC
          Rat Expression Set 230 1369407_PM_at - -
          Rat Expression Set 230 1369407_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903725 - -
          Rat Genome U34 Set U94330_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374220 - -
          RatRef-12 Expression BeadChip ILMN_60409 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline