Search
Search
| Gene Symbol: Wdr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013958.1 | NP_001013980.1 | 3 - 4 | 277 | 57 | |
| XM_006256710.2 | XP_006256772.1 | 3 - 4 | 293 | 57 | ||
| XM_006256712.2 | XP_006256774.1 | 3 - 4 | 404 | 57 | ||
| XM_006256713.2 | XP_006256775.1 | 3 - 4 | 178 | 57 | ||
| XM_006256714.2 | XP_006256776.1 | 3 - 4 | 301 | 57 | ||
| XM_008773071.1 | XP_008771293.1 | 3 - 4 | 191 | 57 | ||
| GenBank mRNA | BC085816.1 | - | 3 - 4 | 277 | 7123612 | 57 |
Set Membership: |
|