Search
Search
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017304.2 | NP_059000.1 | 7 - 8 | 804 | 83 | |
| XM_008764342.1 | XP_008762564.1 | 7 - 8 | 570 | 83 | ||
| XM_008764343.1 | XP_008762565.1 | 7 - 8 | 823 | 83 | ||
| XM_008764344.1 | XP_008762566.1 | 6 - 7 | 527 | 83 | ||
| XM_008764345.1 | XP_008762567.1 | 5 - 6 | 412 | 83 | ||
| GenBank mRNA | X76724.1 | - | 7 - 8 | 914 | 83 | |
Set Membership: |
|