Search
Search
| Gene Symbol: Hrsp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031714.1 | NP_113902.1 | 4 - 5 | 358 | 80 | |
| GenBank mRNA | AF015949.1 | - | 4 - 5 | 358 | 80 | |
| BC078779.1 | - | 4 - 5 | 349 | 7128728 | 80 | |
| D49363.1 | - | 4 - 5 | 328 | 80 | ||
| FQ209529.1 | - | 4 - 5 | 371 | 80 | ||
| FQ209945.1 | - | 3 - 4 | 208 | 80 | ||
| FQ210021.1 | - | 4 - 5 | 363 | 80 | ||
| FQ218425.1 | - | 4 - 5 | 365 | 80 | ||
| FQ218811.1 | - | 4 - 5 | 366 | 80 | ||
| FQ219440.1 | - | 4 - 5 | 365 | 80 | ||
| FQ219496.1 | - | 4 - 5 | 371 | 80 | ||
| X70825.1 | - | 4 - 5 | 326 | 80 | ||
Set Membership: |
|