Search
Search
| Gene Symbol: Rims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052829.1 | NP_439894.1 | 25 - 26 | 4134 | 112 | |
| XM_008766936.1 | XP_008765158.1 | 26 - 27 | 4317 | 112 | ||
| GenBank mRNA | AF007836.1 | - | 25 - 26 | 4134 | 112 | |
| AF199333.1 | - | 26 - 27 | 4320 | 112 | ||
Set Membership: |
|