Search
Search

Slc38a4| Gene Symbol: Slc38a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130748.1 | NP_570104.1 | 12 - 13 | 1299 | 57 | |
| XM_008765663.1 | XP_008763885.1 | 12 - 13 | 1294 | 57 | ||
| GenBank mRNA | AF295535.1 | - | 12 - 13 | 1299 | 57 | |
| BC097292.1 | - | 13 - 14 | 1227 | 7462761 | 57 | |
Set Membership: |
|