Search
Search
| Gene Symbol: Nphs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130828.2 | NP_570841.2 | 6 - 7 | 859 | 80 | |
| GenBank mRNA | AB091379.1 | - | 6 - 7 | 798 | 80 | |
| AF309631.1 | - | 5 - 6 | 537 | 80 | ||
| AY039651.1 | - | 6 - 7 | 859 | 80 | ||
| BC098649.1 | - | 6 - 7 | 841 | 7127895 | 80 | |
Set Membership: |
|