Search
Search
| Gene Symbol: Spg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006987.1 | NP_001006988.1 | 5 - 6 | 665 | 67 | |
| XM_006243251.2 | XP_006243313.1 | 5 - 6 | 696 | 67 | ||
| GenBank mRNA | BC083760.1 | - | 5 - 6 | 665 | 7192933 | 67 |
Set Membership: |
|