Search
Search

RGD1563941| Gene Symbol: RGD1563941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109293.1 | NP_001102763.1 | 1 | 319 | 71 | |
| GenBank mRNA | BC158857.1 | - | 430 | 7930388 | 71 | |
Set Membership: |
|