Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn07318891_s1
          See other CTH GE Assays ›
          Gene Symbol:
          Cth
          Gene Name:
          cystathionine gamma-lyase
          Gene Aliases:
          H
          Chromosome Location:
          Chr.2: 264267029 - 264293041 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn07318891_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cth
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017074.1 NP_058770.1 12 1512 135
          XM_006233536.2 XP_006233598.1 11 1451 135
          GenBank mRNA BC078869.1 - 12 1525 7109746 135
          D17370.1 - 12 1512 135

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cth

          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          negative regulation of cell proliferation
          protein-pyridoxal-5-phosphate linkage via peptidyl-N6-pyridoxal phosphate-L-lysine
          cysteine biosynthetic process
          transsulfuration
          negative regulation of cell growth
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          protein sulfhydration
          homocysteine metabolic process
          positive regulation of NF-kappaB transcription factor activity
          protein homotetramerization
          hydrogen sulfide biosynthetic process
          negative regulation of apoptotic signaling pathway
          cystathionine beta-lyase activity
          cystathionine gamma-lyase activity
          calmodulin binding
          pyridoxal phosphate binding
          L-cystine L-cysteine-lyase (deaminating)
          L-cysteine desulfhydrase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P512874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P512874 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P512874 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P512874 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P512874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82928 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85147 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44919 - -
          Agilent Rat cDNA Microarray (G4105A) 700307241 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10827520 - -
          Affymetrix Rat Gene 1.0 ST Array 10827524 - -
          Affymetrix Rat Gene 1.0 ST Array 10827521 - -
          Affymetrix Rat Gene 1.0 ST Array 10827519 - -
          Affymetrix Rat Gene 1.0 ST Array 10827529 - -
          Affymetrix Rat Gene 1.0 ST Array 10827530 - -
          Affymetrix Rat Gene 1.0 ST Array 10827528 - -
          Affymetrix Rat Gene 1.0 ST Array 10827522 - -
          Affymetrix Rat Gene 1.0 ST Array 10827527 - -
          Affymetrix Rat Gene 1.0 ST Array 10827526 - -
          Affymetrix Rat Gene 1.0 ST Array 10827525 - -
          Affymetrix Rat Gene 1.0 ST Array 10827518 - -
          Affymetrix Rat Gene 1.0 ST Array 10827523 - -
          Affymetrix Rat Gene 1.1 ST Array 10827517 - -
          Rat Genome 230 2.0 Array 1367838_PM_at - -
          Rat Genome 230 2.0 Array 1367838_at - -
          Affymetrix HT Rat Focus Array 1367838_at - -
          Affymetrix HT Rat Focus Array 1367838_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200034246.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034258.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034249.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034254.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034252.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034247.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034248.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034245.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034251.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034250.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034255.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034257.rn.1 TC0200004957.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200034253.rn.1 TC0200004957.rn.1 RTA TC
          Rat Expression Set 230 1367838_PM_at - -
          Rat Expression Set 230 1367838_at - -
          Affymetrix Rat Gene 1.0 ST Array 10827517 - -
          Rat Genome U34 Set D17370_g_at - -
          Rat Genome U34 Set D17370_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370828 - -
          RatRef-12 Expression BeadChip ILMN_67413 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline