Search
Search
| Gene Symbol: Ndrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013923.1 | NP_001013945.1 | 16 | 2417 | 97 | |
| GenBank mRNA | BC078946.1 | - | 16 | 2417 | 7109582 | 97 |
| BC091308.1 | - | 10 | 2106 | 97 | ||
Set Membership: |
|