Search
Search

Tex19.1| Gene Symbol: Tex19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017482.1 | NP_001017482.1 | 3 | 1215 | 102 | |
| GenBank mRNA | BC083916.1 | - | 3 | 1215 | 7134696 | 102 |
Set Membership: |
|