Search
Search

SLC10A2| Gene Symbol: SLC10A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244463.1 | NP_001231392.1 | 3 - 4 | 676 | 123 | |
| GenBank mRNA | AK231499.1 | - | 3 - 4 | 676 | 123 | |
| EF472590.1 | - | 3 - 4 | 340 | 123 | ||
Set Membership: |
|