Search
Search
| Gene Symbol: TOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123205.1 | NP_001116677.1 | 2 | 1355 | 102 | |
| GenBank mRNA | AK234281.1 | - | 2 | 1360 | 102 | |
| AK346102.1 | - | 2 | 1360 | 102 | ||
| AK389488.1 | - | 2 | 1362 | 102 | ||
| AK395727.1 | - | 2 | 1360 | 102 | ||
| AK399312.1 | - | 2 | 1361 | 102 | ||
| EF486515.1 | - | 2 | 1355 | 102 | ||
| EU585747.1 | - | 2 | 1377 | 102 | ||
Set Membership: |
|