Search
Search
| Gene Symbol: SST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009583.1 | NP_001009583.1 | 1 - 2 | 141 | 84 | |
| GenBank mRNA | AK344002.1 | - | 1 - 2 | 247 | 84 | |
| AY609447.1 | - | 1 - 2 | 233 | 84 | ||
Set Membership: |
|