Search
Search

| Gene Symbol: NANOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129971.1 | NP_001123443.1 | 4 | 949 | 90 | |
| GenBank mRNA | FJ882402.1 | - | 1 | 756 | 90 | |
| KM186171.1 | - | 1 | 756 | 90 | ||
| Gene Symbol: NANOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092389.1 | XP_020948048.1 | 1558 | 90 | ||
| XM_021092390.1 | XP_020948049.1 | 1238 | 90 | |||
| GenBank mRNA | DQ447201.1 | - | 4 | 949 | 90 | |
Set Membership: |
|