Search Thermo Fisher Scientific
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185139.1 | NP_001172068.1 | 2 - 3 | 344 | 101 | |
GenBank mRNA | AB529862.1 | - | 2 - 3 | 344 | 101 | |
AK240367.1 | - | 2 - 3 | 455 | 101 | ||
AK347608.1 | - | 2 - 3 | 419 | 101 | ||
AY609633.1 | - | 2 - 3 | 393 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |