Search
Search
| Gene Symbol: PTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244783.1 | NP_001231712.1 | 1 - 2 | 260 | 106 | |
| GenBank mRNA | AK236052.1 | - | 1 - 2 | 260 | 106 | |
| KT861480.1 | - | 1 - 2 | 130 | 106 | ||
Set Membership: |
|