Search
Search

NDUFA4L2| Gene Symbol: NDUFA4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205377.1 | NP_001192306.1 | 1 - 2 | 84 | 108 | |
| XM_013997635.2 | XP_013853089.2 | 5763 | 108 | |||
| GenBank mRNA | AK349810.1 | - | 1 - 2 | 84 | 108 | |
Set Membership: |
|