Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU447172.1 | - | 2 - 3 | 489 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Vitis vinifera Probe spans exons |