Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004982.1 | NP_001004982.1 | 6 - 7 | 846 | 114 | |
XM_012958838.2 | XP_012814292.1 | 6 - 7 | 921 | 114 | ||
XM_012958839.2 | XP_012814293.1 | 6 - 7 | 797 | 114 | ||
GenBank mRNA | BC075533.1 | - | 6 - 7 | 846 | 114 | |
CT030631.1 | - | 6 - 7 | 839 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |