Search
Search

tlnrd1| Gene Symbol: tlnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203654.1 | NP_988985.1 | 1 | 1052 | 81 | |
| GenBank mRNA | BC061331.1 | - | 1 | 1052 | 81 | |
| CT030320.1 | - | 1 | 1124 | 81 | ||
Set Membership: |
|