Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203681.1 | NP_989012.1 | 6 - 7 | 626 | 98 | |
GenBank mRNA | BC061446.1 | - | 6 - 7 | 626 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |