Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011071.1 | NP_001011071.1 | 7 - 8 | 725 | 63 | |
XM_012953486.2 | XP_012808940.1 | 7 - 8 | 706 | 63 | ||
GenBank mRNA | BC084454.1 | - | 7 - 8 | 725 | 63 | |
CR761781.2 | - | 6 - 7 | 633 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |