Search Thermo Fisher Scientific
Gene Symbol: srek1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015999.2 | NP_001015999.1 | 2 - 3 | 135 | 103 | |
GenBank mRNA | BC135675.1 | - | 2 - 3 | 99 | 103 | |
CR848286.2 | - | 2 - 3 | 135 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |