Search
Search

ostc| Gene Symbol: ostc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016656.2 | NP_001016656.1 | 3 - 4 | 272 | 63 | |
| XM_012962599.2 | XP_012818053.1 | 3 - 4 | 400 | 63 | ||
| XM_018093394.1 | XP_017948883.1 | 3 - 4 | 352 | 63 | ||
| GenBank mRNA | BC157801.1 | - | 3 - 4 | 350 | 63 | |
| CR760329.2 | - | 3 - 4 | 272 | 63 | ||
Set Membership: |
|