Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Xt03713968_g1
          See other ELN1 GE Assays ›
          Gene Symbol:
          eln1
          Gene Name:
          tropoelastin 1
          Gene Aliases:
          eln
          Chromosome Location:
          Chr.2: 37685140 - 37763602 on Build UCB_Xtro_10.0
          Species:
          Xenopus tropicalis
          Species Specific ID (Flybase ID):
          -
          Assay ID Xt03713968_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: eln1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001078709.1 NP_001072177.1 51 - 52 3419 64
          XM_012957183.2 XP_012812637.1 54 - 55 3638 64
          XM_012957184.2 XP_012812638.1 53 - 54 3608 64
          XM_012957185.2 XP_012812639.1 53 - 54 3605 64
          XM_012957188.1 XP_012812642.1 54 - 55 3567 64
          XM_012957189.2 XP_012812643.1 52 - 53 3545 64
          XM_012957190.2 XP_012812644.1 52 - 53 3517 64
          XM_012957191.2 XP_012812645.1 52 - 53 3510 64
          XM_012957192.2 XP_012812646.1 52 - 53 3501 64
          XM_012957193.1 XP_012812647.1 52 - 53 3502 64
          XM_012957194.2 XP_012812648.1 52 - 53 3491 64
          XM_012957195.2 XP_012812649.1 51 - 52 3487 64
          XM_012957196.1 XP_012812650.1 52 - 53 3477 64
          XM_012957197.1 XP_012812651.1 52 - 53 3447 64
          XM_012957199.2 XP_012812653.1 50 - 51 3424 64
          XM_012957200.1 XP_012812654.1 50 - 51 3364 64
          XM_012957201.2 XP_012812655.1 50 - 51 3351 64
          XM_012957202.1 XP_012812656.1 50 - 51 3339 64
          XM_012957204.1 XP_012812658.1 50 - 51 3340 64
          XM_012957205.1 XP_012812659.1 50 - 51 3333 64
          XM_012957206.1 XP_012812660.1 50 - 51 3318 64
          XM_012957207.1 XP_012812661.1 48 - 49 3208 64
          XM_012957208.1 XP_012812662.1 48 - 49 3202 64
          XM_012957209.1 XP_012812663.1 46 - 47 3073 64
          XM_012957210.2 XP_012812664.1 46 - 47 3070 64
          XM_012957211.1 XP_012812665.1 46 - 47 3052 64
          XM_018091327.1 XP_017946816.1 48 - 49 3205 64
          GenBank mRNA DQ523561.1 - 51 - 52 3419 64

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons

          Panther Classification:

          Molecular Function -

          structural protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0