Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp26b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079187.2 | NP_001072655.1 | 945 | 67 | ||
GenBank mRNA | BC123940.1 | - | 977 | 67 | ||
BC135551.1 | - | 945 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis |