Search
Search

pck1| Gene Symbol: pck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079568.1 | NP_001073036.1 | 9 - 10 | 1454 | 77 | |
| GenBank mRNA | BC123058.1 | - | 9 - 10 | 1454 | 77 | |
| CR926436.2 | - | 6 - 7 | 77 | |||
| CU075833.1 | - | 9 - 10 | 1501 | 77 | ||
Set Membership: |
|