Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: galr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113883.1 | NP_001107355.1 | 2 - 3 | 853 | 72 | |
GenBank mRNA | BC157202.1 | - | 2 - 3 | 853 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |