Search
Search

cfap58| Gene Symbol: cfap58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126070.1 | NP_001119542.1 | 1384 | 68 | ||
| GenBank mRNA | BC123995.1 | - | 1332 | 68 | ||
| BC166135.1 | - | 1384 | 68 | |||
Set Membership: |
|