Search
Search

rsl24d1| Gene Symbol: rsl24d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126668.1 | NP_001120140.1 | 5 - 6 | 441 | 101 | |
| GenBank mRNA | BC159094.1 | - | 5 - 6 | 441 | 101 | |
| CT025434.2 | - | 5 - 6 | 424 | 101 | ||
Set Membership: |
|