Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nomo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127971.1 | NP_001121443.1 | 15 - 16 | 1870 | 70 | |
GenBank mRNA | BC166195.1 | - | 15 - 16 | 1870 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |