Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: mlo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111701.1 | NP_001105171.1 | 482 | 87 | ||
GenBank mRNA | AY029319.1 | - | 482 | 87 | ||
AY104078.1 | - | 482 | 87 | |||
BT043122.1 | - | 661 | 87 | |||
BT056039.1 | - | 554 | 87 | |||
EU971076.1 | - | 528 | 87 |