Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: tua5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111854.1 | NP_001105324.1 | 1448 | 107 | ||
GenBank mRNA | AY104263.1 | - | 1602 | 107 | ||
L27815.1 | - | 1448 | 107 | |||
U05258.1 | - | 1448 | 107 | |||
X63177.1 | - | 1449 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Zea mays |