Search
Search

BOKAGGCATCCTGTCAGGGAGCGGTCCC[C/T]CCTTGGGACGGTCCCTGGTCATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| BOK - BOK, BCL2 family apoptosis regulator | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_032515.4 | Intron | NP_115904.1 | ||||
| XM_011511696.2 | Intron | XP_011509998.1 | ||||
| XM_011511697.2 | Intron | XP_011509999.1 | ||||
| XM_017004775.1 | Intron | XP_016860264.1 | ||||
| XM_017004776.1 | Intron | XP_016860265.1 | ||||