Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CGCTTTCCGCCGTTACTGGGCGTAT[G/T]GCGTACAGACACGAGGCCGGCGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIWIL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIWIL1 - piwi like RNA-mediated gene silencing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190971.1 | 14 | UTR 5 | NP_001177900.1 | |||
NM_004764.4 | 14 | UTR 5 | NP_004755.2 | |||
XM_011539002.2 | 14 | UTR 5 | XP_011537304.1 | |||
XM_011539003.2 | 14 | Intron | XP_011537305.1 | |||
XM_011539004.2 | 14 | Intron | XP_011537306.1 | |||
XM_011539005.1 | 14 | Intron | XP_011537307.1 | |||
XM_011539006.2 | 14 | Intron | XP_011537308.1 | |||
XM_017020229.1 | 14 | Intron | XP_016875718.1 |