Search
Search

ISL2
SCAPERAAGGGACACTTTCCGACTCGGAGCA[C/G]GCGGCCCTGGAGCACCAGCTCGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ISL2 - ISL LIM homeobox 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_145805.2 | 1178 | Intron | NP_665804.1 | |||
| XM_017022505.1 | 1178 | Missense Mutation | CGC,GGC | R,G 340 | XP_016877994.1 | |
| SCAPER - S-phase cyclin A associated protein in the ER | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||