Search
Search

MIR6784
NMT1
PLCD3TTCCTGGAATTCTTCTCCAGGCCAA[G/A]GCCCCAGGCCTTCAACAACCTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6784 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| MIR6784 - microRNA 6784 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NMT1 - N-myristoyltransferase 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PLCD3 - phospholipase C delta 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_133373.4 | Intron | NP_588614.1 | ||||
| XM_011524253.2 | Intron | XP_011522555.1 | ||||