Search
Search

ZNF329
ZSCAN18ATAACAACTCAGTATTTTTGCTTCA[A/G]CAACTAAAGCAACAAACCACATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF329 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ZNF329 - zinc finger protein 329 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_024620.3 | 3147 | UTR 3 | NP_078896.3 | |||
| XM_006723381.2 | 3147 | UTR 3 | XP_006723444.1 | |||
| XM_006723382.3 | 3147 | UTR 3 | XP_006723445.1 | |||
| XM_006723383.3 | 3147 | UTR 3 | XP_006723446.1 | |||
| XM_006723384.3 | 3147 | UTR 3 | XP_006723447.1 | |||
| XM_011527310.2 | 3147 | UTR 3 | XP_011525612.1 | |||
| XM_011527311.2 | 3147 | UTR 3 | XP_011525613.1 | |||
| XM_011527312.2 | 3147 | UTR 3 | XP_011525614.1 | |||
| XM_011527313.2 | 3147 | UTR 3 | XP_011525615.1 | |||
| XM_011527314.2 | 3147 | UTR 3 | XP_011525616.1 | |||
| XM_011527315.1 | 3147 | UTR 3 | XP_011525617.1 | |||
| XM_011527316.2 | 3147 | UTR 3 | XP_011525618.1 | |||
| XM_017027307.1 | 3147 | UTR 3 | XP_016882796.1 | |||
| XM_017027308.1 | 3147 | UTR 3 | XP_016882797.1 | |||
| XM_017027309.1 | 3147 | UTR 3 | XP_016882798.1 | |||
| XM_017027310.1 | 3147 | UTR 3 | XP_016882799.1 | |||
| XM_017027311.1 | 3147 | UTR 3 | XP_016882800.1 | |||
| ZSCAN18 - zinc finger and SCAN domain containing 18 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||