Search
Search

LOC100288123
MIR1909
REXO1GGGGGTTGGGGATGTGGGCGATCCT[A/C]CTCTTCTCGCCAGGGGCGGAAATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
LOC100288123 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| LOC100288123 - uncharacterized LOC100288123 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR1909 - microRNA 1909 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| REXO1 - RNA exonuclease 1 homolog | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020695.3 | 2323 | Missense Mutation | AGG,AGT | R,S 731 | NP_065746.3 | |
| XM_011528144.1 | 2323 | Missense Mutation | AGG,AGT | R,S 731 | XP_011526446.1 | |
| XM_011528146.2 | 2323 | Missense Mutation | AGG,AGT | R,S 696 | XP_011526448.1 | |
| XM_017027028.1 | 2323 | Missense Mutation | AGG,AGT | R,S 731 | XP_016882517.1 | |
| XM_017027029.1 | 2323 | Missense Mutation | AGG,AGT | R,S 731 | XP_016882518.1 | |
| XM_017027030.1 | 2323 | Missense Mutation | AGG,AGT | R,S 731 | XP_016882519.1 | |