Search
Search

ZNFX1TAAGTATTACCTTCCAGAAACTGGT[A/C]CAAAGCATGATTAGTATAACACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNFX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ZNFX1 - zinc finger NFX1-type containing 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_021035.2 | 1079 | Missense Mutation | GAC,TAC | D,Y 663 | NP_066363.1 | |
| XM_006723836.1 | 1079 | Missense Mutation | GAC,TAC | D,Y 663 | XP_006723899.1 | |
| XM_011528924.1 | 1079 | Missense Mutation | GAC,TAC | D,Y 663 | XP_011527226.1 | |
| XM_017027971.1 | 1079 | UTR 5 | XP_016883460.1 | |||