Search
Search

SNCA
SNCA-AS1AAAAAGTAATTTAATTGAAGATTTG[T/C]GGGAAGATTAAGTCAACATAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| SNCA - synuclein alpha | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000345.3 | Intron | NP_000336.1 | ||||
| NM_001146054.1 | Intron | NP_001139526.1 | ||||
| NM_001146055.1 | Intron | NP_001139527.1 | ||||
| NM_007308.2 | Intron | NP_009292.1 | ||||
| XM_011532203.1 | Intron | XP_011530505.1 | ||||
| XM_011532204.2 | Intron | XP_011530506.1 | ||||
| XM_011532205.2 | Intron | XP_011530507.1 | ||||
| XM_011532206.1 | Intron | XP_011530508.1 | ||||
| XM_011532207.1 | Intron | XP_011530509.1 | ||||
| XM_011532208.2 | Intron | XP_011530510.1 | ||||
| XM_017008562.1 | Intron | XP_016864051.1 | ||||
| XM_017008563.1 | Intron | XP_016864052.1 | ||||
| SNCA-AS1 - SNCA antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||