Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAGAGCTGCCACCAGCAGCAGG[C/T]TCAGACACTGGGCTCCCAGCTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192132 MIM: 604295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1B1 - ATPase H+ transporting V1 subunit B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001692.3 | 54 | UTR 5 | NP_001683.2 | |||
XM_011532907.2 | 54 | Intron | XP_011531209.1 |
ATP6V1B1-AS1 - ATP6V1B1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAX2 - ventral anterior homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |