Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTAAGTTACCCATGCTTTGTGGG[A/G]ACTCAGCCTGCAAAGAATTGAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2F2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2F2 - general transcription factor IIF subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004128.2 | 2046 | Intron | NP_004119.1 | |||
XM_011535052.2 | 2046 | Intron | XP_011533354.1 | |||
XM_011535053.2 | 2046 | Intron | XP_011533355.1 | |||
XM_017020550.1 | 2046 | Intron | XP_016876039.1 | |||
XM_017020551.1 | 2046 | Intron | XP_016876040.1 |
KCTD4 - potassium channel tetramerization domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198404.2 | 2046 | UTR 3 | NP_940686.2 |