Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CAGCCTCTGGGACAAGGCCTCTCTC[C/T]GCAGACCCGGGCCTTACAGGTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FYN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FYN - FYN proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002037.5 | 1731 | UTR 3 | NP_002028.1 | |||
NM_153047.3 | 1731 | UTR 3 | NP_694592.1 | |||
NM_153048.3 | 1731 | UTR 3 | NP_694593.1 | |||
XM_005266890.3 | 1731 | UTR 3 | XP_005266947.1 | |||
XM_005266892.3 | 1731 | UTR 3 | XP_005266949.1 | |||
XM_017010650.1 | 1731 | UTR 3 | XP_016866139.1 | |||
XM_017010651.1 | 1731 | UTR 3 | XP_016866140.1 | |||
XM_017010652.1 | 1731 | UTR 3 | XP_016866141.1 | |||
XM_017010653.1 | 1731 | UTR 3 | XP_016866142.1 | |||
XM_017010654.1 | 1731 | UTR 3 | XP_016866143.1 | |||
XM_017010655.1 | 1731 | Intron | XP_016866144.1 |