Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TTTGAAATTTGGGGAAATTAAGTAG[A/G]CACATGATGAGCTAGCTGAGGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf118 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf118 - chromosome 6 open reading frame 118 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144980.3 | Intron | NP_659417.2 | ||||
XM_005266838.2 | Intron | XP_005266895.1 | ||||
XM_011535509.2 | Intron | XP_011533811.1 | ||||
XM_011535510.2 | Intron | XP_011533812.1 | ||||
XM_011535511.2 | Intron | XP_011533813.1 | ||||
XM_011535512.2 | Intron | XP_011533814.1 | ||||
XM_011535513.2 | Intron | XP_011533815.1 | ||||
XM_017010323.1 | Intron | XP_016865812.1 | ||||
XM_017010324.1 | Intron | XP_016865813.1 |