Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CAAGCCACCTGGAGTTTTCCTTACC[C/T]GAATTTCTGCCCCTTCACGCCGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLS2 - glutaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280796.1 | 1614 | Missense Mutation | CAG,CGG | Q,R 207 | NP_001267725.1 | |
NM_001280797.1 | 1614 | Missense Mutation | CAG,CGG | Q,R 218 | NP_001267726.1 | |
NM_001280798.1 | 1614 | Missense Mutation | CAG,CGG | Q,R 218 | NP_001267727.1 | |
NM_013267.3 | 1614 | Missense Mutation | CAG,CGG | Q,R 483 | NP_037399.2 | |
XM_005268797.1 | 1614 | Missense Mutation | CAG,CGG | Q,R 447 | XP_005268854.1 | |
XM_017019180.1 | 1614 | Missense Mutation | CAG,CGG | Q,R 218 | XP_016874669.1 |
SPRYD4 - SPRY domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |