Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AATCTTCTTCCTCTTCCTCGGTTCG[C/T]TTGTGTTTTAAAAACAAGTCAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS10 - regulator of G-protein signaling 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005339.1 | 540 | Silent Mutation | AAA,AAG | K,K 156 | NP_001005339.1 | |
NM_002925.3 | 540 | Silent Mutation | AAA,AAG | K,K 142 | NP_002916.1 | |
XM_017016492.1 | 540 | Silent Mutation | AAA,AAG | K,K 94 | XP_016871981.1 | |
XM_017016493.1 | 540 | Silent Mutation | AAA,AAG | K,K 108 | XP_016871982.1 |